Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122505.4 | NP_568480.2 | 10 - 11 | 1062 | 101 | |
NM_180747.2 | NP_851078.1 | 10 - 11 | 932 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |