Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343982.1 | NP_001331385.1 | 9 - 10 | 1096 | 133 | |
NM_122560.2 | NP_568488.2 | 9 - 10 | 1038 | 133 | ||
NM_180749.4 | NP_851080.1 | 9 - 10 | 1096 | 133 | ||
NM_180750.2 | NP_851081.1 | 9 - 10 | 1038 | 133 | ||
GenBank mRNA | AK318672.1 | - | 9 - 10 | 1030 | 133 | |
BT001097.1 | - | 9 - 10 | 934 | 133 | ||
BT001113.1 | - | 9 - 10 | 934 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |