Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203501.1 | NP_001190430.1 | 4 - 5 | 530 | 77 | |
NM_001344186.1 | NP_001330326.1 | 2 - 3 | 1473 | 77 | ||
NM_001344187.1 | NP_001330327.1 | 2 - 3 | 292 | 77 | ||
NM_001344188.1 | NP_001330328.1 | 1 - 2 | 207 | 77 | ||
NM_123096.3 | NP_198553.2 | 3 - 4 | 516 | 77 | ||
NM_180770.2 | NP_851101.1 | 3 - 4 | 587 | 77 | ||
GenBank mRNA | AK318988.1 | - | 5 - 6 | 534 | 77 | |
AY093216.1 | - | 3 - 4 | 516 | 77 | ||
BT008856.1 | - | 2 - 3 | 368 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |