Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203501.1 | NP_001190430.1 | 5 - 6 | 604 | 82 | |
NM_001344186.1 | NP_001330326.1 | 3 - 4 | 1547 | 82 | ||
NM_001344187.1 | NP_001330327.1 | 3 - 4 | 366 | 82 | ||
NM_001344188.1 | NP_001330328.1 | 2 - 3 | 281 | 82 | ||
NM_180770.2 | NP_851101.1 | 4 - 5 | 661 | 82 | ||
GenBank mRNA | AK318988.1 | - | 6 - 7 | 608 | 82 | |
AY093216.1 | - | 4 - 5 | 590 | 82 | ||
BT008856.1 | - | 3 - 4 | 442 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Arabidopsis thaliana Probe spans exons |