Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344582.1 | NP_001332492.1 | 1 - 2 | 350 | 74 | |
NM_123873.3 | NP_199318.2 | 1 - 2 | 350 | 74 | ||
NM_180802.2 | NP_851133.1 | 1 - 2 | 518 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |