Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124156.5 | NP_199593.2 | 11 - 12 | 1923 | 60 | |
NM_180820.2 | NP_851151.1 | 11 - 12 | 1944 | 60 | ||
GenBank mRNA | AY158083.1 | - | 11 - 12 | 1594 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |