Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124198.4 | NP_568694.2 | 9 - 10 | 1099 | 68 | |
NM_180823.3 | NP_851154.1 | 8 - 9 | 1169 | 68 | ||
GenBank mRNA | AF364059.1 | - | 9 - 10 | 968 | 68 | |
AK317237.1 | - | 9 - 10 | 872 | 68 | ||
AY059736.1 | - | 7 - 8 | 837 | 68 | ||
AY091271.1 | - | 7 - 8 | 766 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |