Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036960.2 | NP_001032037.1 | 7 - 8 | 618 | 102 | |
NM_001344808.1 | NP_001331840.1 | 7 - 8 | 790 | 102 | ||
NM_124265.5 | NP_568704.2 | 6 - 7 | 1485 | 102 | ||
NM_180826.4 | NP_851157.1 | 6 - 7 | 635 | 102 | ||
GenBank mRNA | AF062590.1 | - | 6 - 7 | 515 | 102 | |
AF062591.1 | - | 6 - 7 | 734 | 102 | ||
AY079026.1 | - | 6 - 7 | 620 | 102 | ||
AY136455.1 | - | 6 - 7 | 523 | 102 | ||
BT002594.1 | - | 5 - 6 | 422 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |