Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125941.1 | NP_001119413.1 | 2 - 3 | 509 | 115 | |
NM_124502.3 | NP_199936.2 | 3 - 4 | 501 | 115 | ||
NM_180837.3 | NP_851168.1 | 3 - 4 | 593 | 115 | ||
GenBank mRNA | AB053171.1 | - | 3 - 4 | 438 | 115 | |
AY140086.1 | - | 3 - 4 | 450 | 115 | ||
BT008388.1 | - | 2 - 3 | 190 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |