Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125941.1 | NP_001119413.1 | 6 - 7 | 885 | 99 | |
NM_124502.3 | NP_199936.2 | 7 - 8 | 877 | 99 | ||
NM_180837.3 | NP_851168.1 | 7 - 8 | 969 | 99 | ||
GenBank mRNA | AB053171.1 | - | 7 - 8 | 814 | 99 | |
AY140086.1 | - | 7 - 8 | 826 | 99 | ||
BT008388.1 | - | 6 - 7 | 566 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |