Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085279.1 | NP_001078748.1 | 6 - 7 | 621 | 80 | |
NM_001085280.1 | NP_001078749.1 | 5 - 6 | 526 | 80 | ||
NM_001345033.1 | NP_001332707.1 | 5 - 6 | 652 | 80 | ||
NM_124691.3 | NP_568785.1 | 6 - 7 | 658 | 80 | ||
NM_180847.2 | NP_851178.1 | 5 - 6 | 718 | 80 | ||
NM_180848.3 | NP_851179.1 | 5 - 6 | 705 | 80 | ||
GenBank mRNA | AY040013.1 | - | 5 - 6 | 684 | 80 | |
AY079367.1 | - | 4 - 5 | 440 | 80 | ||
BT000742.1 | - | 5 - 6 | 669 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |