Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345288.1 | NP_001318824.1 | 5 - 6 | 1440 | 130 | |
NM_001345289.1 | NP_001330355.1 | 4 - 5 | 1079 | 130 | ||
NM_001345290.1 | NP_001330356.1 | 3 - 4 | 1429 | 130 | ||
NM_125199.2 | NP_568874.2 | 4 - 5 | 899 | 130 | ||
NM_180879.2 | NP_851210.1 | 4 - 5 | 1256 | 130 | ||
NM_180880.2 | NP_851211.1 | 4 - 5 | 1252 | 130 | ||
NM_180881.3 | NP_851212.2 | 3 - 4 | 1037 | 130 | ||
GenBank mRNA | AF053941.2 | - | 3 - 4 | 794 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |