Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345288.1 | NP_001318824.1 | 9 - 10 | 2040 | 80 | |
NM_001345289.1 | NP_001330355.1 | 8 - 9 | 1679 | 80 | ||
NM_001345290.1 | NP_001330356.1 | 7 - 8 | 2029 | 80 | ||
NM_125199.2 | NP_568874.2 | 8 - 9 | 1499 | 80 | ||
NM_180879.2 | NP_851210.1 | 8 - 9 | 1856 | 80 | ||
NM_180880.2 | NP_851211.1 | 8 - 9 | 1852 | 80 | ||
NM_180881.3 | NP_851212.2 | 7 - 8 | 1637 | 80 | ||
GenBank mRNA | AF053941.2 | - | 7 - 8 | 1394 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |