Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIL1-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203652.1 | NP_001190581.1 | 8 - 9 | 1384 | 74 | |
NM_125457.3 | NP_568926.2 | 10 - 11 | 1576 | 74 | ||
NM_180903.4 | NP_851234.1 | 10 - 11 | 1636 | 74 | ||
GenBank mRNA | BT001994.1 | - | 10 - 11 | 1574 | 74 | |
BT008359.1 | - | 10 - 11 | 1531 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |