Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125495.2 | NP_568932.2 | 5 - 6 | 1248 | 81 | |
NM_180905.3 | NP_851236.1 | 5 - 6 | 1311 | 81 | ||
GenBank mRNA | AK226952.1 | - | 5 - 6 | 1254 | 81 | |
AY065413.1 | - | 5 - 6 | 1224 | 81 | ||
AY096551.1 | - | 5 - 6 | 1149 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |