Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345460.1 | NP_001330163.1 | 1 - 2 | 311 | 152 | |
NM_125518.5 | NP_200933.2 | 2 - 3 | 370 | 152 | ||
NM_180907.3 | NP_851238.1 | 2 - 3 | 372 | 152 | ||
GenBank mRNA | AK317449.1 | - | 2 - 3 | 362 | 152 | |
AY099736.1 | - | 2 - 3 | 293 | 152 | ||
BT002603.1 | - | 1 - 2 | 212 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |