Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125534.4 | NP_200949.1 | 3 - 4 | 641 | 76 | |
NM_180909.3 | NP_851240.1 | 3 - 4 | 749 | 76 | ||
GenBank mRNA | AF015274.1 | - | 3 - 4 | 473 | 76 | |
AY045855.1 | - | 3 - 4 | 500 | 76 | ||
AY091381.1 | - | 3 - 4 | 368 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |