Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125534.4 | NP_200949.1 | 5 - 6 | 833 | 74 | |
NM_180909.3 | NP_851240.1 | 5 - 6 | 941 | 74 | ||
GenBank mRNA | AF015274.1 | - | 5 - 6 | 665 | 74 | |
AY045855.1 | - | 5 - 6 | 692 | 74 | ||
AY091381.1 | - | 5 - 6 | 560 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |