Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345521.1 | NP_001318862.1 | 1 - 2 | 398 | 72 | |
NM_001345522.1 | NP_001332748.1 | 1 - 2 | 451 | 72 | ||
NM_001345523.1 | NP_001332747.1 | 1 - 2 | 398 | 72 | ||
NM_001345524.1 | NP_001332749.1 | 1 - 2 | 343 | 72 | ||
NM_125603.3 | NP_568950.2 | 1 - 2 | 451 | 72 | ||
NM_180915.1 | NP_851246.1 | 1 - 2 | 258 | 72 | ||
GenBank mRNA | AK228933.1 | - | 1 - 2 | 232 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |