Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125820.3 | NP_201229.1 | 2 - 3 | 951 | 96 | |
NM_180931.2 | NP_851262.1 | 2 - 3 | 951 | 96 | ||
GenBank mRNA | AY086951.1 | - | 2 - 3 | 771 | 96 | |
AY322527.1 | - | 2 - 3 | 735 | 96 | ||
BT002395.1 | - | 2 - 3 | 769 | 96 | ||
BT010372.1 | - | 2 - 3 | 735 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |