Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125893.5 | NP_201299.2 | 5 - 6 | 1155 | 86 | |
NM_180940.2 | NP_851271.1 | 5 - 6 | 1155 | 86 | ||
GenBank mRNA | AY059877.1 | - | 5 - 6 | 1138 | 86 | |
AY064044.1 | - | 5 - 6 | 1114 | 86 | ||
AY096382.1 | - | 5 - 6 | 993 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |