Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037070.1 | NP_001032147.1 | 2 - 3 | 400 | 99 | |
NM_001037071.1 | NP_001032148.1 | 2 - 3 | 307 | 99 | ||
NM_001037072.2 | NP_001032149.1 | 3 - 4 | 327 | 99 | ||
NM_001345675.1 | NP_001318881.1 | 2 - 3 | 637 | 99 | ||
NM_125919.3 | NP_201324.1 | 1 - 2 | 1097 | 99 | ||
NM_180942.3 | NP_851273.1 | 2 - 3 | 710 | 99 | ||
GenBank mRNA | AK317142.1 | - | 2 - 3 | 308 | 99 | |
AK317488.1 | - | 3 - 4 | 328 | 99 | ||
AY099593.1 | - | 2 - 3 | 426 | 99 | ||
AY128838.1 | - | 1 - 2 | 178 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |