Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZWI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203699.1 | NP_001190628.1 | 8 - 9 | 1639 | 87 | |
NM_125990.4 | NP_569022.2 | 8 - 9 | 1746 | 87 | ||
NM_180945.2 | NP_851276.1 | 8 - 9 | 1691 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |