Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115005.5 | NP_190714.2 | 11 - 12 | 1241 | 132 | |
GenBank mRNA | AF266459.1 | - | 10 - 11 | 867 | 132 | |
AF360267.1 | - | 11 - 12 | 961 | 132 | ||
AY040078.1 | - | 10 - 11 | 867 | 132 | ||
AY227250.1 | - | 10 - 11 | 867 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |