Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115037.8 | NP_190746.2 | 1 - 2 | 675 | 82 | |
GenBank mRNA | AY090347.1 | - | 1 - 2 | 312 | 82 | |
AY143837.1 | - | 1 - 2 | 205 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |