Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115141.3 | NP_190849.1 | 5 - 6 | 1196 | 101 | |
GenBank mRNA | AF492667.1 | - | 5 - 6 | 1103 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |