Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161202.1 | NP_001154674.1 | 5 - 6 | 441 | 103 | |
NM_115148.4 | NP_190856.1 | 4 - 5 | 488 | 103 | ||
GenBank mRNA | AF360197.1 | - | 4 - 5 | 283 | 103 | |
AK227030.1 | - | 4 - 5 | 312 | 103 | ||
AK317341.1 | - | 4 - 5 | 311 | 103 | ||
AY045666.1 | - | 4 - 5 | 311 | 103 | ||
AY057576.1 | - | 4 - 5 | 339 | 103 | ||
AY060525.1 | - | 4 - 5 | 257 | 103 | ||
AY070718.1 | - | 4 - 5 | 330 | 103 | ||
AY091403.1 | - | 4 - 5 | 257 | 103 | ||
AY125492.1 | - | 4 - 5 | 309 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |