Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115154.3 | NP_566975.1 | 10 - 11 | 983 | 122 | |
NM_202691.1 | NP_974420.1 | 10 - 11 | 942 | 122 | ||
GenBank mRNA | AF256535.1 | - | 10 - 11 | 891 | 122 | |
AK227189.1 | - | 10 - 11 | 849 | 122 | ||
AY064964.1 | - | 10 - 11 | 951 | 122 | ||
AY133650.1 | - | 10 - 11 | 767 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |