Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115172.4 | NP_190880.1 | 4 - 5 | 412 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |