Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339651.1 | NP_001326764.1 | 1 - 2 | 140 | 75 | |
NM_115274.3 | NP_190982.1 | 2 - 3 | 406 | 75 | ||
GenBank mRNA | AF360155.1 | - | 2 - 3 | 233 | 75 | |
AY084773.1 | - | 2 - 3 | 222 | 75 | ||
AY113884.1 | - | 1 - 2 | 104 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |