Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIP37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115277.5 | NP_190985.1 | 4 - 5 | 491 | 135 | |
GenBank mRNA | AF084570.1 | - | 4 - 5 | 373 | 135 | |
AY063940.1 | - | 4 - 5 | 398 | 135 | ||
AY113943.1 | - | 4 - 5 | 307 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |