Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203156.1 | NP_001190085.1 | 3 - 4 | 280 | 131 | |
NM_115288.8 | NP_190996.3 | 3 - 4 | 404 | 131 | ||
GenBank mRNA | AB440793.1 | - | 2 - 3 | 142 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |