Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115322.4 | NP_567005.2 | 2 - 3 | 1060 | 110 | |
GenBank mRNA | AF462827.1 | - | 2 - 3 | 754 | 110 | |
AY142006.1 | - | 2 - 3 | 728 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |