Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084818.1 | NP_001078287.1 | 6 - 7 | 658 | 108 | |
NM_001339701.1 | NP_001327724.1 | 5 - 6 | 576 | 108 | ||
NM_115341.3 | NP_567008.1 | 6 - 7 | 658 | 108 | ||
GenBank mRNA | AB007766.2 | - | 6 - 7 | 568 | 108 | |
AK316975.1 | - | 6 - 7 | 609 | 108 | ||
BT002860.1 | - | 6 - 7 | 563 | 108 | ||
BT004392.1 | - | 6 - 7 | 473 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |