Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATVPS33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161207.1 | NP_001154679.1 | 20 - 21 | 1667 | 66 | |
NM_115343.4 | NP_567009.1 | 19 - 20 | 1732 | 66 | ||
GenBank mRNA | AF357527.1 | - | 19 - 20 | 1552 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |