Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATVPS33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161207.1 | NP_001154679.1 | 7 - 8 | 728 | 94 | |
NM_115343.4 | NP_567009.1 | 7 - 8 | 841 | 94 | ||
GenBank mRNA | AF357527.1 | - | 7 - 8 | 661 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |