Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339702.1 | NP_001326426.1 | 7 - 8 | 1349 | 72 | |
NM_001339703.1 | NP_001326427.1 | 7 - 8 | 1311 | 72 | ||
NM_115344.4 | NP_191047.3 | 7 - 8 | 1223 | 72 | ||
GenBank mRNA | AB290928.1 | - | 7 - 8 | 1238 | 72 | |
EU257694.1 | - | 7 - 8 | 1223 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |