Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115384.4 | NP_567017.2 | 10 - 11 | 1225 | 130 | |
GenBank mRNA | AM493720.1 | - | 10 - 11 | 1004 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |