Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115391.5 | NP_191093.1 | 5 - 6 | 617 | 109 | |
GenBank mRNA | AF419553.1 | - | 5 - 6 | 576 | 109 | |
AY087663.1 | - | 5 - 6 | 512 | 109 | ||
AY097338.1 | - | 5 - 6 | 500 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |