Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115398.3 | NP_191100.1 | 9 - 10 | 1675 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |