Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115410.3 | NP_191112.1 | 6 - 7 | 701 | 65 | |
GenBank mRNA | AF386995.1 | - | 6 - 7 | 671 | 65 | |
AK317055.1 | - | 6 - 7 | 688 | 65 | ||
BT006305.1 | - | 5 - 6 | 533 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |