Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115462.4 | NP_567031.1 | 13 - 14 | 2405 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |