Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115462.4 | NP_567031.1 | 8 - 9 | 1968 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |