Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339805.1 | NP_001326776.1 | 7 - 8 | 1582 | 124 | |
NM_115529.2 | NP_191229.1 | 7 - 8 | 1519 | 124 | ||
GenBank mRNA | DQ056628.1 | - | 7 - 8 | 1422 | 124 | |
EU280151.1 | - | 7 - 8 | 1485 | 124 | ||
EU280152.1 | - | 7 - 8 | 1422 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |