Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339835.1 | NP_001327450.1 | 13 - 14 | 1538 | 129 | |
NM_001339836.1 | NP_001327449.1 | 13 - 14 | 1510 | 129 | ||
NM_115558.6 | NP_191258.2 | 13 - 14 | 1538 | 129 | ||
GenBank mRNA | AY069919.1 | - | 13 - 14 | 1425 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |