Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115567.3 | NP_191267.1 | 3 - 4 | 700 | 121 | |
GenBank mRNA | AK118958.1 | - | 3 - 4 | 568 | 121 | |
BT005410.1 | - | 3 - 4 | 547 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |