Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339858.1 | NP_001319779.1 | 7 - 8 | 853 | 119 | |
NM_001339859.1 | NP_001325644.1 | 7 - 8 | 853 | 119 | ||
NM_001339860.1 | NP_001325645.1 | 7 - 8 | 1618 | 119 | ||
NM_001339861.1 | NP_001325651.1 | 7 - 8 | 1672 | 119 | ||
NM_001339862.1 | NP_001325648.1 | 7 - 8 | 1287 | 119 | ||
NM_001339863.1 | NP_001325647.1 | 7 - 8 | 1325 | 119 | ||
NM_001339864.1 | NP_001325650.1 | 7 - 8 | 833 | 119 | ||
NM_001339865.1 | NP_001325649.1 | 6 - 7 | 989 | 119 | ||
NM_001339866.1 | NP_001325646.1 | 6 - 7 | 1027 | 119 | ||
NM_115601.3 | NP_567048.1 | 7 - 8 | 891 | 119 | ||
GenBank mRNA | AK318626.1 | - | 6 - 7 | 844 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |