Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT3G57470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203189.1 | NP_001190118.1 | 14 - 15 | 1535 | 85 | |
NM_001339871.1 | NP_001319782.1 | 14 - 15 | 1447 | 85 | ||
NM_001339872.1 | NP_001326452.1 | 14 - 15 | 1539 | 85 | ||
NM_001339873.1 | NP_001326451.1 | 14 - 15 | 1539 | 85 | ||
NM_001339874.1 | NP_001326450.1 | 14 - 15 | 1535 | 85 | ||
NM_115607.4 | NP_567049.3 | 14 - 15 | 1443 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |