Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT3G57470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203189.1 | NP_001190118.1 | 22 - 23 | 2636 | 88 | |
NM_001339871.1 | NP_001319782.1 | 22 - 23 | 2548 | 88 | ||
NM_001339872.1 | NP_001326452.1 | 22 - 23 | 2640 | 88 | ||
NM_001339873.1 | NP_001326451.1 | 22 - 23 | 2640 | 88 | ||
NM_001339874.1 | NP_001326450.1 | 22 - 23 | 2636 | 88 | ||
NM_115607.4 | NP_567049.3 | 22 - 23 | 2544 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |