Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT3G57470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203189.1 | NP_001190118.1 | 3 - 4 | 507 | 150 | |
NM_001339871.1 | NP_001319782.1 | 3 - 4 | 419 | 150 | ||
NM_001339872.1 | NP_001326452.1 | 3 - 4 | 511 | 150 | ||
NM_001339873.1 | NP_001326451.1 | 3 - 4 | 511 | 150 | ||
NM_001339874.1 | NP_001326450.1 | 3 - 4 | 507 | 150 | ||
NM_115607.4 | NP_567049.3 | 3 - 4 | 415 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |